Learn More
Meso Scale Discovery V-PLEX NHP IL-12/IL-23p40 Kit
SDP
Supplier:  Meso Scale Discovery K156RJD2
Encompass
The Meso Scale Discovery V-PLEX NHP IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of natural human IL-12 and IL-23 in serum plasma and culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG read buffer wash buffer blocker solutions calibrators and controls The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81113