Learn More
Meso Scale Discovery V-PLEX NHP IL-16 Kit 25Plate
SDP
Supplier:  Meso Scale Discovery K156RED4
Encompass
The Meso Scale Discovery V-PLEX NHP IL-16 Kit is a sensitive immunoassay for measuring human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s platforms providing quantitative read-outs in pg/mL This assay aids in studying inflammatory diseases and immune function
Catalog No. 50-000-80809
Only null left
Add to Cart
Request a Quote