Learn More
Meso Scale Discovery V-PLEX NHP IL-2 Kit
SDP
Supplier:  Meso Scale Discovery K156QQD4
Encompass
The V-PLEX NHP IL-2 Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for quantifying natural human interleukin 2 (IL-2) in cell culture supernatants serum and plasma It includes a 96-well plate with a monoclonal antibody for IL-2 a detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 06 pg/mL with a range of 0 06-1500 pg/mL requiring 50 L of sample and delivering results in 3 5 hours It is specific for human IL-2 with no cross-reactivity to other cytokines ideal for measuring IL-2 from T cell activation
Catalog No. 50-000-79020
Add to Cart
Request a Quote