Learn More
Meso Scale Discovery V-PLEX NHP IL-7 Kit 25Plate
SDP
Supplier:  Meso Scale Discovery K156RCD4
Encompass
The Meso Scale Discovery V-PLEX NHP IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a detection range of 0 06-1000 pg/mL and a lower limit of detection of 0 03 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-80829
Add to Cart
Request a Quote