Learn More
Meso Scale Discovery V-PLEX NHP MCP-4 Kit
SDP
Supplier:  Meso Scale Discovery K156NOD2
Encompass
The V-PLEX NHP MCP-4 Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of 4 neurodegenerative biomarkers in human serum plasma and CSF samples The kit contains a 96-well plate coated with capture antibodies for the biomarkers A 42 t-tau p-tau and -synuclein It utilizes electrochemiluminescent detection and provides high sensitivity a broad dynamic range and requires only a small sample volume The kit includes all necessary reagents and can process up to 80 samples in duplicate It is intended for research use only
Catalog No. 50-000-81145