Learn More
Meso Scale Discovery V-PLEX NHP TNF-SY Kit
SDP
Supplier:  Meso Scale Discovery K156RGD1
Encompass
The V-PLEX NHP TNF-beta Kit from Meso Scale Discovery is a highly sensitive immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human TNF-beta in serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators and reagents required to perform the assay The kit has a detection range of 0 64-2500 pg/mL and exhibits minimal cross-reactivity with other cytokines This assay allows high-throughput screening and provides accurate reproducible results with as little as 12 5 L sample volume
Catalog No. 50-000-79967