Learn More
Meso Scale Discovery V-PLEX NHP VEGF Kit 25Plate
SDP
Supplier:  Meso Scale Discovery K156RHD4
Encompass
The V-PLEX NHP VEGF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of VEGF-A in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies for VEGF-A detection antibodies calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 3 2-10 000 pg/mL and takes 3 5 hours to complete It utilizes electrochemiluminescent detection for high sensitivity and wide dynamic range The kit includes reagents for one 96-well plate which can test 40 samples in duplicate
Catalog No. 50-000-79941
Only null left
Add to Cart
Request a Quote