Learn More
Meso Scale Discovery V-PLEX Plus GIP active Kit
SDP
Supplier:  Meso Scale Discovery K150T5G2
Encompass
The Meso Scale Discovery V-PLEX Plus GIP Active Kit is a 96-well plate assay for measuring active GLP-1 levels in serum plasma and cell culture supernatants This electrochemiluminescence immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification of active GLP-1 down to 0 15 pg/mL It includes a 96-well plate with a GLP-1 capture antibody a detection antibody with an MSD SULFO-TAG and calibrators and controls for a standard curve The assay enables high-throughput analysis of active GLP-1 in just 3 5 hours
Catalog No. 50-000-82288