Learn More
Meso Scale Discovery V-PLEX Plus GLP-1 Active Kit
SDP
Supplier:  Meso Scale Discovery K1503OG1
Encompass
The Meso Scale Discovery V-PLEX Plus GLP-1 Active Kit measures active GLP-1 in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection It includes a 96-well plate with a GLP-1 capture antibody a detection antibody with an MSD SULFO-TAG and calibrators and controls for a standard curve With a 2-hour incubation and minimal sample volume it enables high-throughput quantification of active GLP-1
Catalog No. 50-000-78769