Learn More
Meso Scale Discovery V-PLEX Plus Human Ab42 Kit
SDP
Supplier:  Meso Scale Discovery K151LBG1
Encompass
The V-PLEX Plus Human A 42 Kit from Meso Scale Discovery is an immunoassay that quantitatively measures amyloid beta 1-42 (A 42) in human cerebrospinal fluid (CSF) serum and plasma It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a plate coated with a monoclonal antibody to human A 42 a ruthenylated detection antibody calibrators diluent read buffer and wash buffer It allows rapid measurement of A 42 levels a biomarker associated with Alzheimer s disease The small sample size required makes this kit useful for studies where sample volumes are limited
Catalog No. 50-000-82277