Learn More
Meso Scale Discovery V-PLEX Plus Human Eotaxin Kit
SDP
Supplier:  Meso Scale Discovery K151NSG2
Encompass
The Meso Scale Discovery V-PLEX Human Eotaxin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human Eotaxin in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions The assay has a sensitivity of 0 06 pg/mL and a detection range of 0 06-500 pg/mL The total assay time is 3 5 hours This assay allows for the accurate quantitation of human Eotaxin levels
Catalog No. 50-000-82473