Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery V-PLEX Plus Human IFN-γ Kit, 25 plates, Single-analyte, 96-well, Sensitivity 0.37 pg/mL, Dynamic Range 0.37 - 938 pg/mL, Serum/Plasma/Urine/Cell Culture
SDP

Supplier:  Meso Scale Discovery K151QOG4

Encompass

The MESO SCALE DISCOVERY V-PLEX Plus Human IFN-γ Kit is a sensitive immunoassay for quantifying human Interferon-gamma (IFN-γ) levels in various biological samples. This kit is analytically validated for consistent performance, making it suitable for research in cytokines & chemokines, as well as immunology and inflammation applications.

  • Analyte: IFN-γ
  • Species: Human
  • Assay platform: V-PLEX Plus
  • Assay format: Single-analyte
  • Application areas: Cytokines & chemokines, Immunology/Inflammation
  • Plate type: 96-well
  • Sensitivity (LLOD): 0.37 pg/mL
  • Dynamic range: 0.37 - 938 pg/mL
  • Sample types: Serum, Plasma, Urine, Cell culture supernatant
  • Quantity: 25 plates
  • Usage statement: For research use only
  • Capture antibody: Mouse monoclonal
  • Detection antibody: Mouse monoclonal

Catalog No. NC3727495


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.