Learn More
Meso Scale Discovery V-PLEX Plus Human IFN-y Kit
SDP
Supplier:  Meso Scale Discovery K151QOG1
Encompass
The V-PLEX Plus Human IFN gamma Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) for measuring human interferon gamma (IFN-) in various samples It features MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators diluent wash buffer read buffer and plate seals It detects as low as 0 05 pg/mL and up to 2500 pg/mL in cell culture supernatants
Catalog No. 50-000-81669