Learn More
Meso Scale Discovery V-PLEX Plus Human IL-10 Kit
SDP
Supplier:  Meso Scale Discovery K151QUG4
Encompass
The Meso Scale Discovery Human IL-10 Assay Kit is designed to quantitatively measure human interleukin 10 (IL-10) in a variety of sample types such as serum plasma cell culture supernatants and other biological fluids The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay can detect IL-10 concentrations as low as 0 2 pg/mL and has a 7-log dynamic range The total assay time is less than 4 hours for 96-well plates This assay kit offers a rapid sensitive and robust method for measuring human IL-10 levels
Catalog No. 50-000-81677
Add to Cart
Request a Quote