Learn More
Meso Scale Discovery V-PLEX Plus Human IL-10 Kit
SDP
Supplier:  Meso Scale Discovery K151QUG2
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-10 Kit is a highly sensitive immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human interleukin 10 (IL-10) in cell culture supernatants serum plasma and other biological fluids The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-1500 pg/mL and requires 25 L sample size The total assay time is 3 5 hours This ECL-based assay provides superior sensitivity a wide dynamic range and requires less sample volume compared to ELISA
Catalog No. 50-000-82191