Learn More
Meso Scale Discovery V-PLEX Plus Human IL-10 Kit
SDP
Supplier:  Meso Scale Discovery K151QUG1
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-1500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SULFO-TAG labels and MULTI-ARRAY technology to provide sensitive and reproducible quantitation of IL-6
Catalog No. 50-000-82322