Learn More
Meso Scale Discovery V-PLEX Plus Human IL-15 Kit
SDP
Supplier:  Meso Scale Discovery K151RDG2
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-15 a detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a detection range of 3 2-10 000 pg/mL and exhibits no cross-reactivity with other cytokines The kit takes approximately 3 hours to run and only requires a minimal sample volume of 50 L
Catalog No. 50-000-79835