Learn More
Meso Scale Discovery V-PLEX Plus Human IL-16 Kit
SDP
Supplier:  Meso Scale Discovery K151REG1
Encompass
The Meso Scale Discovery V-PLEX Human IL-16 Kit is a sensitive immunoassay for measuring human interleukin 16 (IL-16) in various biological fluids It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and minimal sample volume The kit includes a 96-well plate with capture and detection antibodies buffers and a lyophilized calibrator It detects IL-16 from 0 05-1000 pg/mL requiring 2-fold sample dilutions
Catalog No. 50-000-78761