Learn More
Meso Scale Discovery V-PLEX Plus Human IL-1a Kit
SDP
Supplier:  Meso Scale Discovery K151RBG4
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-1alpha Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 1 alpha (IL-1alpha) in cell culture supernatants serum plasma and other biological fluids This kit uses MSD s proprietary V-PLEX technology based on electrochemiluminescence providing high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL
Catalog No. 50-000-78840
Add to Cart
Request a Quote