Learn More
Meso Scale Discovery V-PLEX Plus Human IL-2 Kit
SDP
Supplier:  Meso Scale Discovery K151QQG1
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 2 (IL-2) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody to human IL-2 a vial of SULFO-TAG labeled anti-human IL-2 detection antibody assay and sample diluents a plate reader and a software package for data analysis The assay has a sensitivity of 0 06 pg/mL and a broad dynamic range from 0 06-1500 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of IL-2 levels in biological samples
Catalog No. 50-000-78789