Learn More
Meso Scale Discovery V-PLEX Plus Human IL-5 Kit
SDP
Supplier:  Meso Scale Discovery K151QSG1
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 5 (IL-5) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-5 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 08 pg/mL and a 7-point standard curve from 2500 to 2 4 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 tests in duplicate
Catalog No. 50-000-80586