Learn More
Meso Scale Discovery V-PLEX Plus Human IL-6 Kit
SDP
Supplier:  Meso Scale Discovery K151QXG1
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 0 06-1500 pg/mL and exhibits high sensitivity specificity and reproducibility for quantifying IL-6 levels The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78794