Learn More
Meso Scale Discovery V-PLEX Plus Human MCP-1 Kit
SDP
Supplier:  Meso Scale Discovery K151NNG2
Encompass
The V-PLEX Plus Human MCP-1 Kit is an immunoassay that quantitatively measures human MCP-1 in various biological samples such as serum plasma and cell culture supernatants This kit utilizes electrochemiluminescence (ECL) technology and contains all necessary reagents to complete the assay The kit has high sensitivity and wide dynamic range It requires low sample volume and has high reproducibility This assay can be used to study chemokine biology and inflammation
Catalog No. 50-000-79269