Learn More
Meso Scale Discovery V-PLEX Plus Human MCP-4 Kit
SDP
Supplier:  Meso Scale Discovery K151NOG4
Encompass
The Meso Scale Discovery V-PLEX Human MCP-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human Monocyte Chemotactic Protein 4 (MCP-4) in cell culture supernates serum and plasma The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to run This quantitative immunoassay allows sensitive and accurate measurement of MCP-4 levels
Catalog No. 50-000-81197
Add to Cart
Request a Quote