Learn More
Meso Scale Discovery V-PLEX Plus Human MDC Kit
SDP
Supplier:  Meso Scale Discovery K151NPG1
Encompass
The V-PLEX Plus Human MDC Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous detection and quantification of 10 human cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IFN- TNF- IP-10 and MCP-1) from cell culture supernatants serum plasma and other biological samples The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls
Catalog No. 50-000-81198