Learn More
Meso Scale Discovery V-PLEX Plus Human MIP-1a Kit
SDP
Supplier:  Meso Scale Discovery K151NQG2
Encompass
The V-PLEX Human MIP-1 alpha Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human MIP-1 alpha in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL
Catalog No. 50-000-79923