Learn More
Meso Scale Discovery V-PLEX Plus Human SAA Kit
SDP
Supplier:  Meso Scale Discovery K151SSG1
Encompass
The Meso Scale Discovery V-PLEX Plus Human SAA Kit is an electrochemiluminescent immunoassay for the quantitative measurement of serum amyloid A (SAA) in human serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a range from 0 49 pg/mL to 400 pg/mL and takes approximately 3 5 hours to complete This product can aid in the evaluation of inflammatory conditions and immune responses
Catalog No. 50-000-82222