Learn More
Meso Scale Discovery V-PLEX Plus Human TARC Kit
SDP
Supplier:  Meso Scale Discovery K151NTG1
Encompass
The V-PLEX Plus Human TARC Kit from Meso Scale Discovery is a multiplex immunoassay measuring TARC/CCL17 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive quantitation of TARC with a detection range of 0 3-2500 pg/mL and a 25 L sample volume The kit includes a 96-well plate detection antibody Read Buffer Calibrator Diluent and lyophilized SULFO-TAG labeled detection antibody enabling high-throughput measurement of TARC levels to aid in understanding Th2-mediated immune responses
Catalog No. 50-000-81201