Learn More
Meso Scale Discovery V-PLEX Plus Human Tie-2 Kit
SDP
Supplier:  Meso Scale Discovery K151KWG1
Encompass
The Meso Scale Discovery V-PLEX Plus Human TIE-2 Kit is a multiplex immunoassay that quantitatively measures human TIE-2 in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay Results are analyzed on MSD s benchtop MESO QuickPlex SQ 120 instrument This kit allows for the accurate quantification of human TIE-2 levels
Catalog No. 50-000-82237