Learn More
Meso Scale Discovery V-PLEX Plus Human Tie-2 Kit
SDP
Supplier:  Meso Scale Discovery K151KWG4
Encompass
The Meso Scale Discovery V-PLEX Plus Human TIE-2 Kit is a multiplex immunoassay that quantitatively measures human TIE-2 in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators diluent wash buffer read buffer and plate seals This allows for the accurate quantification of human TIE-2 levels in approximately 3 5 hours
Catalog No. 50-000-82350
Only null left
Add to Cart
Request a Quote