Learn More
Meso Scale Discovery V-PLEX Plus Human VCAM-1 Kit
SDP
Supplier:  Meso Scale Discovery K151SRG1
Encompass
The V-PLEX Plus Human VCAM-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human VCAM-1 in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The V-PLEX kit can reliably quantify VCAM-1 across a range of 0 53 - 2500 pg/mL This product would be well-suited for research applications focused on analyzing VCAM-1 levels as a biomarker of endothelial dysfunction and inflammation
Catalog No. 50-000-79161