Learn More
Meso Scale Discovery V-PLEX Plus Human VCAM-1 Kit
SDP
Supplier:  Meso Scale Discovery K151SRG4
Encompass
The Meso Scale Discovery V-PLEX Plus Human VCAM-1 Kit is a multiplex immunoassay that quantitatively measures human VCAM-1 in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators controls 10X diluent read buffer and plate seals The assay has a typical lower limit of detection of 0 23 ng/mL and an upper limit of quantification of 10 000 ng/mL
Catalog No. 50-000-81299
Add to Cart
Request a Quote