Learn More
Meso Scale Discovery V-PLEX Plus Human VEGF-D Kit
SDP
Supplier:  Meso Scale Discovery K151LUG2
Encompass
The Meso Scale Discovery V-PLEX Plus Human VEGF-D Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human VEGF-D in cell culture supernates serum plasma and other biological fluids It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a rapid ultra-sensitive and cost-effective assay with high reproducibility This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and diluent The assay has a typical lower limit of detection of 3 5 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-78673