Learn More
Meso Scale Discovery V-PLEX Plus Human VEGF-D Kit
SDP
Supplier:  Meso Scale Discovery K151LUG4
Encompass
The Meso Scale Discovery V-PLEX Plus Human VEGF-D Kit is an electrochemiluminescent immunoassay for quantifying human VEGF-D in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive specific and rapid measurement with low background The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a 25 L sample covering a dynamic range from picogram to nanogram levels
Catalog No. 50-000-78675
Add to Cart
Request a Quote