Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-12p70 Kit
SDP
Supplier:  Meso Scale Discovery K152QVG2
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-12p70 Kit is an electrochemiluminescence immunoassay for the quantitative measurement of mouse interleukin 12 subunit p70 (IL-12p70) in cell culture supernatants serum plasma and other biological fluids This kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples to generate a standard curve and validate assay performance The V-PLEX assay utilizes patented MULTI-ARRAY technology to provide sensitive accurate and reproducible quantification of mouse IL-12p70 levels with a broad dynamic range and requires just 50-100 L of sample
Catalog No. 50-000-78439