Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-5 Kit
SDP
Supplier:  Meso Scale Discovery K152QSG2
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 5 (IL-5) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and includes reagents for testing 2 x 96-well plates This quantitative immunoassay allows sensitive and accurate measurement of mouse IL-5 with a testing time of 3 5 hours
Catalog No. 50-000-78706