Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-6 Kit
SDP
Supplier:  Meso Scale Discovery K152QXG2
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 6 (IL-6) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for cell culture supernatants The total assay time is 3 5 hours This is a sensitive accurate and reproducible kit for quantifying mouse IL-6 levels
Catalog No. 50-000-78647