Learn More
Meso Scale Discovery V-PLEX Plus Mouse IP-10 Kit
SDP
Supplier:  Meso Scale Discovery K152NVG2
Encompass
The V-PLEX Plus Mouse IP-10 Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for quantifying IP-10 in mouse serum plasma and cell culture supernatants It includes a 96-well plate with IP-10 capture antibody detection antibody calibrator diluent wash buffer read buffer and calibrator Sensitivity is 1 53 pg/mL with a quantitation limit of 10 000 pg/mL The kit uses electrochemiluminescence for high sensitivity and rapid results testing 40 samples in duplicate
Catalog No. 50-000-80039