Learn More
Meso Scale Discovery V-PLEX Plus Mouse KC/GRO Kit
SDP
Supplier:  Meso Scale Discovery K152QTG2
Encompass
The V-PLEX Plus Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Streptavidin-HRP Using electrochemiluminescence and MSD s MULTI-ARRAY technology it provides sensitive results from small samples aiding in cytokine profiling and inflammation studies
Catalog No. 50-000-80434