Learn More
Meso Scale Discovery V-PLEX Plus Mouse MCP-1 Kit
SDP
Supplier:  Meso Scale Discovery K152NNG4
Encompass
The V-PLEX Plus Mouse MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse MCP-1 in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a lyophilized mouse MCP-1 calibrator The assay sensitivity is 1 3 pg/mL with an upper quantitation limit of 10 000 pg/mL allowing up to 94 measurements across two plates This kit enables high-throughput sensitive and accurate quantification of mouse MCP-1
Catalog No. 50-000-81499
Only null left
Add to Cart
Request a Quote