Learn More
Meso Scale Discovery V-PLEX Plus Mouse MIP-1 Kit
SDP
Supplier:  Meso Scale Discovery K152NQG4
Encompass
The V-PLEX Plus Mouse MIP-1 alpha Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures mouse MIP-1 alpha in cell culture supernatants serum plasma and other biological fluids The kit uses electrochemiluminescence technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve The assay has a detection range of 0 03-2500 pg/mL and requires only 25 L of sample
Catalog No. 50-000-81382
Add to Cart
Request a Quote