Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-10 Kit
SDP
Supplier:  Meso Scale Discovery K156QUG4
Encompass
Meso Scale Discovery offers an IL-10 assay kit for the quantitative measurement of IL-10 in non-human primate samples such as serum plasma and cell culture supernatants The assay uses MSD s electrochemiluminescence detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay can be used to support non-human primate research in areas such as infectious disease immunology neuroscience and drug development
Catalog No. 50-000-79949
Add to Cart
Request a Quote