Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-16 Kit
SDP
Supplier:  Meso Scale Discovery K156REG1
Encompass
The V-PLEX Plus NHP IL-16 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of IL-16 in non-human primate serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents optimized for use with non-human primate samples The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for IL-16
Catalog No. 50-000-79916