Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-17A Kit
SDP
Supplier:  Meso Scale Discovery K156RFG1
Encompass
The Meso Scale Discovery IL-17A Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 17A (IL-17A) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with IL-17A capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to run the assay The kit can detect IL-17A in the range of 0 3-2500 pg/mL
Catalog No. 50-000-78544