Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-1SY Kit
SDP
Supplier:  Meso Scale Discovery K156QPG1
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-1 Kit is a sensitive immunoassay for measuring natural human primate interleukin-1 beta (IL-1 ) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity wide dynamic range and minimal matrix interference in a 96-well plate With detection limits from 0 04 to 10 000 pg/mL it quantifies IL-1 accurately The assay requires 50 L of sample per well and is read on MSD s SECTOR Imager in 3 5 hours
Catalog No. 50-000-79945