Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-2 Kit
SDP
Supplier:  Meso Scale Discovery K156QQG2
Encompass
The V-PLEX Plus NHP IL-2 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of natural human IL-2 in serum plasma and cell culture supernatants The kit includes a plate coated with a monoclonal antibody to human IL-2 a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 06 pg/mL and a range of 0 06-1500 pg/mL It requires 2 5 hours to complete and is designed for use on Meso Scale Discovery instruments The kit provides materials for one 96-well plate which can test up to 40 samples in duplicate
Catalog No. 50-000-79973