Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-5 Kit
SDP
Supplier:  Meso Scale Discovery K156QSG2
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 5 (IL-5) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-5 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 03 pg/mL and a broad dynamic range from 0 03-500 pg/mL This immunoassay allows for the accurate and sensitive quantification of IL-5 levels
Catalog No. 50-000-81249