Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-6 Kit
SDP
Supplier:  Meso Scale Discovery K156QXG1
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-6 Kit is an electrochemiluminescent immunoassay for quantifying human interleukin 6 (IL-6) in serum plasma and culture supernatants It includes a 96-well plate with a monoclonal antibody to IL-6 a detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 06 pg/mL with a range of 0 06-1500 pg/mL It takes about 3 5 hours to run and has enough reagents for one 96-well plate
Catalog No. 50-000-78227