Learn More
Meso Scale Discovery V-PLEX Plus NHP MIP-1SY Kit
SDP
Supplier:  Meso Scale Discovery K156NRG1
Encompass
The Meso Scale Discovery MIP-1 beta Kit is a singleplex immunoassay that uses electrochemiluminescence (ECL) detection technology for the quantitative measurement of MIP-1 beta in serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibody detection antibody labeled with an ECL compound calibrators and buffers The assay has a detection range of 3 2-10 000 pg/mL and takes approximately 3 5 hours to complete It uses a small sample volume of 50 L and has high sensitivity and wide dynamic range The kit includes all necessary reagents and can quantify natural and recombinant human MIP-1 beta
Catalog No. 50-000-80774