Learn More
Meso Scale Discovery V-PLEX Plus NHP VEGF Kit
SDP
Supplier:  Meso Scale Discovery K156RHG1
Encompass
The V-PLEX Plus NHP VEGF Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of VEGF in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies specific for VEGF detection antibodies conjugated to an electrochemiluminescent label calibrator solutions diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and exhibits minimal cross-reactivity to related analytes The kit leverages MSD s proprietary MULTI-ARRAY technology and provides high sensitivity a wide dynamic range and requires a low sample volume of 50 L
Catalog No. 50-000-81390